Skip to Content
Merck
All Photos(1)

Documents

MES-SA

95051030, human uterus, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MES-SA, 95051030

biological source

human uterus

growth mode

Adherent

karyotype

Modal no. 45, monosomic forms of chromosomes 5, 6 and 7

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human uterus sarcoma

Cell Line Description

A human uterine sarcoma derived in 1980 from a tumour obtained at the time of a hysterectomy from a 56 year old Caucasian female. The cells form tumours in athymic nude mice, readily form colonies in soft agar and have a reported doubling time of 22-24 hours. The cells are sensitive to a number of chemotherapeutic agents including doxorubicin, dactinomycin, mitomycin C, taxol and bleomycin. They are resistant to vinblastine dacarbazine, cisplatin, melphalan, vincristine methotrexate and etoposide. The multi drug-resistant cell line MES-SA/Dx5 was established from MES-SA cells grown in the presence of doxorubicin.

Application

Tumourigenicity and chemotherapeutic agent studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 11,13
D16S539: 11,12
D5S818: 13
D7S820: 7,11
THO1: 6
TPOX: 8,11
vWA: 18

Culture Medium

McCoy′s 5a + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:6 to 1:8 i.e. seeding at 1-4 x 10,000 cells/cm2 using 0.03% EDTA; 5% CO2; 37°C. Do not use trypsin.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service