Skip to Content
Merck
All Photos(1)

Documents

HTC

93120108, rat liver, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HTC, 93120108

biological source

rat liver

growth mode

Adherent

karyotype

2n = 66, hypertriploid

morphology

Epithelial

products

Not specified

receptors

Dexamethasone phosphate; hydrocortisone

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat hepatoma

Cell Line Description

Derived from a rat ascites tumour, which in turn was derived from a solid hepatoma induced in male Buffalo rats by feeding them 0.04% N,N′-2,7fluorenyle-bis-2,2,2-trifluoroacetamide for 12.4 months.

Application

Tumourigenicity and enzymatic studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service