Skip to Content
Merck
All Photos(1)

Documents

MPC-11

91031103, mouse blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

MPC-11, 91031103

biological source

mouse blood

growth mode

Suspension

karyotype

Modal no. 67-70

morphology

Lymphoblast

products

Immunoglobulin G (IgG2b), half molecules, light chain dimers, free light chains

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/C Myeloma

Cell Line Description

Established from the Merwin plasma cell tumour-II carried in BALB/c mice. Resembles the parent tumor in that they both produce fully assembled gamma globulin molecules (IgG2b) as well as half molecules, light chain dimers and free light chains. The MPC-11 produces 5-6 picograms of IgG2b per cell per minute.

Culture Medium

DMEM + 2mM Glutamine + 20% Horse Serum (HS).

Subculture Routine

Maintain cultures between 2-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service