Skip to Content
Merck
All Photos(1)

Key Documents

SW 48

89012702, human colon, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human colon
growth mode:
Adherent
karyotype:
2n = 46
morphology:
Epithelial
products:
Carcinoembryonic antigen (CEA)
receptors:
Not specified
relevant disease(s):
cancer

Product Name

SW 48, 89012702

biological source

human colon

growth mode

Adherent

karyotype

2n = 46

morphology

Epithelial

products

Carcinoembryonic antigen (CEA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon adenocarcinoma

Cell Line Description

The cells produce a low level of CEA: 0.6ng/1,000,000 cells in 10 days. Cells are tumorigenic in nude mice. Established from a grade IV ulcerating tumour encircling the bowel of an 82 year old Caucasian female. HLA = A32, A33, B27, B35

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 9,10
D13S317: 11,12
D16S539: 11,13
D5S818: 10,14
D7S820: 9,10
THO1: 6,9.3
TPOX: 8
vWA: 18,20

Culture Medium

L15 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; No CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service