Skip to Content
Merck
All Photos(1)

Documents

Hep2 (HeLa derivative) Cell Line human

86030501, human cervix, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Hep2 (HeLa derivative) Cell Line human, 86030501

biological source

human cervix

description

Human Black, cervix carcinoma

growth mode

Adherent

karyotype

Modal no. 14

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human cervix carcinoma

Cell Line Description

Originated from tumours produced in irradiated-cortisonised weanling rats after injecting with epidermoid carcinoma tissue from the larynx of a 56 year old male. This cell line was found to be indistinguishable from HeLa by STR PCR DNA profiling. Therefore, the cell line should be considered as derived from HeLa. Ethnicity: Black.

Application

Human Hep2 (HeLa derivative) cell line has been used for virus and tumorigenicity studies.

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. After thawing the first subculture may take 7-10 days.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service