Skip to Content
Merck
All Photos(1)

Key Documents

BMSIPD960025BS

Sigma-Aldrich

BeadBug Stainless Steel grinding medium, gringding beads, grinding medium

bead diam. 25 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NA.82
Pricing and availability is not currently available.

material

stainless steel

manufacturer/tradename

Benchmark Scientific IPD9600-25BS

bead diam.

25 mm

compatibility

50mL stainless steel grinding jars

Looking for similar products? Visit Product Comparison Guide

General description

Pack of loose 25mm Stainless Steel grinding balls for use with tube homogenizers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service