Skip to Content
Merck
All Photos(2)

Documents

AV34401

Sigma-Aldrich

Anti-IER2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ETR101, Anti-Immediate early response 2, Anti-MGC111472, Anti-MGC15265

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

24 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IER2(9592)

General description

IER2 is an immediate early response gene that regulates FGF signaling and patterning in zebrafish. It is also known to enhance tumor cell invasion, and hence functions as a prognostic marker for colorectal cancer.
Rabbit Anti-IER2 antibody recognizes human IER2.

Immunogen

Synthetic peptide directed towards the N terminal region of human IER2

Application

Rabbit Anti-IER2 antibody can be used for western blot applications at a concentration of 1 μg/ml. It can also be used for IHC applications at 4-8 μg/ml, using paraffin-embedded tissues.

Sequence

Synthetic peptide located within the following region: LYLSAKVEALEPEVSLPAALPSDPRLHPPREAESTAETATPDGEHPFPEP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Neeb et al.
Oncogene, 31(33), 3796-3806 (2011-11-29)
Here, we report unbiased screens for genes expressed in metastatic tumor cells that are associated with cell motility. These screens identified Ier2, an immediate early gene of unknown function, as potentially having a role in tumor cell motility and metastasis.
Sung-Kook Hong et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(7), 2230-2235 (2009-01-24)
Establishment of left-right asymmetry in vertebrates requires nodal, Wnt-PCP and FGF signaling and involves ciliogenesis in a laterality organ. Effector genes through which FGF signaling affects laterality have not been described. We isolated the zebrafish ier2 and fibp1 genes as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service