Key Documents
Z414042
Sigma-Aldrich
Aldrich® enlarging adapter
female joint: ST/NS 55/50, male joint: ST/NS 24/40
Synonym(s):
glass adapter, glass adapters
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Pricing and availability is not currently available.
Joint
female joint: ST/NS55/50
male joint: ST/NS24/40
Related Categories
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)