Skip to Content
Merck
All Photos(1)

Documents

Y0001365

Etoposide for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Etoposide, 4′-Demethylepipodophyllotoxin 9-(4,6-O-ethylidene-β-D-glucopyranoside), VP-16-213

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H32O13
CAS Number:
Molecular Weight:
588.56
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

etoposide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(cc(OC)c1O)[C@H]2[C@@H]3[C@H](COC3=O)[C@H](O[C@@H]4O[C@@H]5CO[C@@H](C)O[C@H]5[C@H](O)[C@H]4O)c6cc7OCOc7cc26

InChI

1S/C29H32O13/c1-11-36-9-20-27(40-11)24(31)25(32)29(41-20)42-26-14-7-17-16(38-10-39-17)6-13(14)21(22-15(26)8-37-28(22)33)12-4-18(34-2)23(30)19(5-12)35-3/h4-7,11,15,20-22,24-27,29-32H,8-10H2,1-3H3/t11-,15+,20-,21-,22+,24-,25-,26-,27-,29+/m1/s1

InChI key

VJJPUSNTGOMMGY-MRVIYFEKSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Etoposide for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Etoposide is an antitumor agent that complexes with topoisomerase II and DNA to enhance double-strand and single-strand cleavage of DNA and reversibly inhibit religation. Blocks the cell cycle in in S-phase and G2-phase of the cell cycle; induces apoptosis in normal and tumor cell lines; inhibits synthesis of the oncoprotein Mdm2 and induces apoptosis in tumor lines that overexpress Mdm2.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abukar Ali et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(1), 107-116 (2014-10-11)
Permanent reduction in joint function is a severe postinfectious complication in patients with Staphylococcus aureus septic arthritis. We undertook this study to determine whether this reduction in joint function might be caused by persistent joint inflammation after the adequate eradication
Sachiko Ezoe
International journal of environmental research and public health, 9(7), 2444-2453 (2012-08-02)
Etoposide is an anticancer agent, which is successfully and extensively used in treatments for various types of cancers in children and adults. However, due to the increases in survival and overall cure rate of cancer patients, interest has arisen on
Olga V Leontieva et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(24), 8832-8837 (2014-06-04)
During cell cycle arrest caused by contact inhibition (CI), cells do not undergo senescence, thus resuming proliferation after replating. The mechanism of senescence avoidance during CI is unknown. Recently, it was demonstrated that the senescence program, namely conversion from cell
Nouhoum Sako et al.
The American journal of pathology, 184(11), 2899-2907 (2014-09-10)
Extranodal natural killer-T-cell lymphoma (NKTCL) of nasal type is a malignant disorder of cytotoxic lymphocytes of natural killer or more rarely T cells, associated with clonal Epstein-Barr virus infection. NKTCL is an aggressive neoplasm with very poor prognosis. Although the
P B Knudsen et al.
Leukemia, 28(6), 1289-1298 (2013-11-28)
Chronic lymphocytic leukemia (CLL) is an incurable malignancy of mature B cells. One of the major challenges in treatment of CLL is the achievement of a complete remission to prevent relapse of disease originating from cells within lymphoid tissues and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service