Skip to Content
Merck
All Photos(1)

Key Documents

Y0001262

Flucytosine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Fluorocytosine, 4-amino-5-fluoro-2(1H)-pyrimidinone, Flucytosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4FN3O
CAS Number:
Molecular Weight:
129.09
Beilstein:
127285
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

flucytosine

manufacturer/tradename

EDQM

mp

298-300 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC1=NC(=O)NC=C1F

InChI

1S/C4H4FN3O/c5-2-1-7-4(9)8-3(2)6/h1H,(H3,6,7,8,9)

InChI key

XRECTZIEBJDKEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Flucytosine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Nucleoside analog that has antifungal activities. 5-FC is deaminated by cytosine deaminase to product 5-fluorouracil, resulting in RNA miscoding. 5-Fluorocytosine inhibits DNA and RNA synthesis and interferes with ribosomal protein synthesis.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheshta Sharma et al.
Mycoses, 57(9), 544-552 (2014-04-05)
We investigated the prevalence of vulvovaginal candidiasis due to C. africana in an STD clinic in India and analysed the genetic relatedness of these C. africana isolates with those outside India. A total of 283 germ-tube-positive yeasts were identified by
Ya-li Yang et al.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases, 26, 154-161 (2014-07-27)
Cryptococcosis is a severe fungal infection with a high mortality rate among solid-organ transplant recipients. Today, China is among the countries performing the most kidney transplants worldwide, however data on the association of cryptococcosis with kidney transplantation in mainland China
Sebastian A Mikolajczak et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 22(9), 1707-1715 (2014-05-16)
Immunization with live-attenuated Plasmodium sporozoites completely protects against malaria infection. Genetic engineering offers a versatile platform to create live-attenuated sporozoite vaccine candidates. We previously generated a genetically attenuated parasite (GAP) by deleting the P52 and P36 genes in the NF54
Hedda Luise Koehling et al.
Journal of clinical microbiology, 53(1), 255-261 (2014-11-14)
Candidemia is an important cause of morbidity and mortality in immunosuppressed patients. Candida isolates must be cultivated, identified, and tested for susceptibility. We compared the performance of a new colorimetric broth microdilution panel (SensiQuattro Candida EU) for antifungal susceptibility testing
Plinio Trabasso et al.
Mycopathologia, 179(1-2), 53-62 (2014-12-08)
Candida parapsilosis complex (CPC) is the third Candida species isolated in blood cultures of patients from our Hospital, following C. albicans and C. tropicalis. From 2006 to 2010, the median annual distribution of CPC was 8 cases/year. Records of 36

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service