Skip to Content
Merck
All Photos(1)

Key Documents

NIST8568

Nitrogen and Oxygen Isotopes in Nitrate (USGS34)

NIST®SRM®

Synonym(s):

Potassium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
MDL number:

isotopic purity

 15N
 17O
 18O

packaging

pkg of 0.9 g

manufacturer/tradename

NIST®

mp

334 °C (lit.)

format

matrix material

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Other Notes

Reference/Informational value is provided for the following analytes.
Nitrogen-15, Oxygen-18, Oxygen-17

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service