Skip to Content
Merck
All Photos(1)

Documents

NIST694

Phosphate rock

NIST® SRM® 694, Western

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 90 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

A single unit of SRM694 consists of 90 g of <150 µm (100 mesh) powdered material. For more information, please refer to the COA and SDS.
SRM 694_cert
SRM 694_SDS

Application

Phosphate rock Standard Reference Material (SRM) is intended for validating analytical methods and calibrating instruments used in the analysis of phosphate rock, fertilizers, and similar materials.

Features and Benefits

NIST certified values for constituents are provided and are expressed in concentration.

Other Notes

Certified for the analytes listed below.
Aluminium oxide (Al2O3), Calcium oxide (CaO), Cadmium oxide (CdO), Flurine (F), Iron oxide (Fe2O3), Potassium oxide (K2O), ,Magnesium oxide (MgO), Manganese oxide (MnO), Sodium oxide (Na2O), Phosphorus pentoxide (P2O5), Silicon dioxide (SiO2), Uranium (U), Vanadium pentoxide (V2O5)

Reference/Informational value is provided for the following analytes.
Chromiun oxide (Cr2O3), Titanium dioxide (TiO2), Zinc oxide (ZnO)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service