NIST3108
Cadmium (Cd) standard solution
NIST® SRM® 3108
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 x 10 mL
manufacturer/tradename
NIST®
format
single component solution
General description
SRM 3108_cert
SRM 3108_SDS
Application
Features and Benefits
- The certified value of the SRM was determined by calculating the mass fraction values of Cadmium which were obtained through two methods:
- Gravimetric preparation.
- Inductively coupled plasma optical emission spectrometry (ICP-OES) .
- The certified value is metrologically traceable to the SI unit of mass fraction, expressed in units of milligrams per gram.
- The NIST certificate is provided with expiration certificate, storage, handling, use, and maintenance instructions.
Other Notes
Cadmium (Cd)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 1B - Repr. 1B - Skin Corr. 1B - STOT RE 2
Target Organs
Kidney,Bone
Supplementary Hazards
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service