Skip to Content
Merck
All Photos(1)

Key Documents

NIST1879A

Respirable cristobalite

NIST® SRM® 1879a, quantitative X-ray powder diffraction standard

Synonym(s):

Cristobalite

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

form

powder

packaging

pkg of 5 g

manufacturer/tradename

NIST®

SMILES string

[Si](=O)=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

General description

Application


  • Inhalable Porous Particles for Lung Drug Delivery: Research on inhalable porous particles of cristobalite demonstrates significant advancements in respiratory drug delivery systems, specifically targeting tuberculosis. These findings emphasize the crucial role of respirable cristobalite in enhancing the efficacy of pulmonary treatments (Campos Pacheco et al., 2024).

  • Impact of Crystalline Silica on Pulmonary Health: A study exploring the effects of crystalline silica-induced inflammation and autoimmunity highlights the importance of respirable cristobalite in understanding occupational health risks and developing protective strategies against lung diseases (Heine et al., 2024).

  • Workplace Exposure to Respirable Dust: Analysis of respirable dust and cristobalite concentrations in workers at a brick kiln in Nepal provides insights into occupational exposure and its health implications, aiding in the development of better industrial hygiene practices (Beard et al., 2024).

  • Asbestos and Cristobalite Analysis: Research on distinguishing asbestos from its nonasbestiform analogues, including cristobalite, through FTIR and multivariate data analysis, underscores the significance of high-purity cristobalite in environmental and material safety studies (Lee et al., 2024).

  • Occupational Exposure Limits Comparison: A comparative study on the occupational exposure limits of coal dust, which includes cristobalite, between China and other countries, highlights the global variations and the need for standardized safety protocols in industrial settings (Li and Zhu, 2024).

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service