Skip to Content
Merck
All Photos(1)

Key Documents

D2962005

Dosulepin impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(E)-N,N-Dimethyl-3-(5-oxidodibenzo[b,e]thiepin-11(6H)-ylidene)-1-propanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NOS
CAS Number:
Molecular Weight:
311.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dosulepin, dothiepin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dosulepin impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Lact. - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K P Maguire et al.
Journal of affective disorders, 4(1), 41-48 (1982-03-01)
Ten patients suffering from primary depressive illness were treated with 150 mg/d of dothiepin for 4 weeks. Blood and plasma concentrations of dothiepin and two metabolites, dothiepin S-oxide and northiaden were measured by gas chromatography-mass spectrometry. Severity of depression was
A Fulton et al.
Journal of affective disorders, 4(3), 261-269 (1982-09-01)
The affinities of dothiepin and its principal metabolites northiaden, dothiepin sulphoxide and northiaden sulphoxide for [3H]imipramine binding sites in the rat cortical homogenates, and for [3H]spiperone and [3H]serotonin receptor sites in preparations from the rat frontal cortex and hippocampus were
K P Maguire et al.
Journal of chromatography, 222(3), 399-408 (1981-03-13)
A method for the simultaneous measurement of dothiepin and two of its major metabolites, northiaden and dothiepin S-oxide in both plasma and whole blood is described. The method involves the use of gas chromatography-mass fragmentography. It is selective, sensitive (1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service