Skip to Content
Merck
All Photos(1)

Key Documents

90479

Supelco

Glyphosate-2-13C,15N

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C13CH215NHCH2PO3H2
CAS Number:
Molecular Weight:
171.06
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥99% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

mp

230 °C (dec.) (lit.)

application(s)

agriculture
environmental

format

neat

mass shift

M+2

SMILES string

OC(=O)[13CH2][15NH]CP(O)(O)=O

InChI

1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)/i1+1,4+1

InChI key

XDDAORKBJWWYJS-VFZPYAPFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Glyphosate is one of the most used herbicides, especially in agriculture applications such as weed and vegetation control.

Application

Glyphosate-2-13C,15N may be used as a reference standard for the determination of glyphosate-2-13C,15N in saline natural water samples by high-performance liquid chromatography coupled to tandem mass spectrometry (HPLC-MS/MS). It may also be used as an internal standard for the analysis of glyphosate, aminomethylphosphonic acid and glufosinate in water samples by online solid-phase extraction (SPE) followed by HPLC/MS.

Analysis Note

13C enrichment ≥99 atom% based upon labeld precursor
15N enrichment ≥98 atom% based upon labeld precursor

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of an Enrichment and LC-MS/MS Method for the Analysis of Glyphosate and Aminomethylphosphonic Acid (AMPA) in Saline Natural Water Samples without Derivatization
Pupke D, et al.
Journal of Chromatography and Separation Techniques, 7(338), 2-2 (2016)
Methods of analysis by the US geological survey organic geochemistry research group-determination of glyphosate, aminomethylphosphonic acid, and glufosinate in water using online solid-phase extraction and high-performance liquid chromatography/mass spectrometry
Lee EA, et al.
Journal of Chromatography and Separation Techniques, 7(338), 2-2 (2002)
Lene Nørby Nielsen et al.
Environmental pollution (Barking, Essex : 1987), 233, 364-376 (2017-11-03)
Recently, concerns have been raised that residues of glyphosate-based herbicides may interfere with the homeostasis of the intestinal bacterial community and thereby affect the health of humans or animals. The biochemical pathway for aromatic amino acid synthesis (Shikimate pathway), which

Articles

Glyphosate and related compounds are measured in oatmeal and infant cereal using ion-exchange polymer-based particles for HPLC and SPE. Low level detection was obtained.

Glyphosate and related compounds are measured in oatmeal and infant cereal using ion-exchange polymer-based particles for HPLC and SPE. Low level detection was obtained.

Glyphosate and related compounds are measured in oatmeal and infant cereal using ion-exchange polymer-based particles for HPLC and SPE. Low level detection was obtained.

Glyphosate and related compounds are measured in oatmeal and infant cereal using ion-exchange polymer-based particles for HPLC and SPE. Low level detection was obtained.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service