Skip to Content
Merck
All Photos(1)

Key Documents

45459

Supelco

Dipropetryn

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21N5S
CAS Number:
Molecular Weight:
255.38
Beilstein:
614796
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCSc1nc(NC(C)C)nc(NC(C)C)n1

InChI

1S/C11H21N5S/c1-6-17-11-15-9(12-7(2)3)14-10(16-11)13-8(4)5/h7-8H,6H2,1-5H3,(H2,12,13,14,15,16)

InChI key

NPWMZOGDXOFZIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinpei Li et al.
Journal of separation science, 39(23), 4585-4591 (2016-11-01)
A novel and simple supported ionic-liquid-based solid-phase extraction method for the determination of triazine herbicides in rice was developed. Glass slides were functionalized by an ionic liquid, 1-carboxyethyl-3-methylimidazolium chloride, and were used for the simultaneous extraction of seven triazine herbicides
Shanshan Wang et al.
Journal of hazardous materials, 367, 686-693 (2019-01-18)
A novel dual-template molecularly imprinted polymer (DMIP) was prepared with atrazine and prometryn as the template and applied as a class-specific adsorbent for simultaneously selective solid-phase extraction of seventeen triazine herbicides and metabolites from complex matrices. For comparison, a non-imprinted
Jishi Wang et al.
Journal of chromatography. A, 1521, 10-18 (2017-09-25)
In this research, the manual two-step QuEChERS approach has been streamlined and automated into a one-step method using a cleanup tube fitted within an extraction tube. A novel automatic QuEChERS combination have been developed to simplify the QuEChERS procedures and
Hui Xu et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 32(2), 183-188 (2016-02-11)
The pneumatic nebulization gas-solid microextraction device fascinating us is because it directly atomized organic samples to cross a solid-phase microextraction (SPME) cartridge without any pretreatment. In this work, both the spray chamber and SPME column of the extraction device were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service