Skip to Content
Merck
All Photos(1)

Key Documents

34088

Supelco

Nitroxinil

VETRANAL®, analytical standard

Synonym(s):

4-Hydroxy-3-iodo-5-nitrobenzonitrile, Nitroxynil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H3IN2O3
CAS Number:
Molecular Weight:
290.01
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Oc1c(I)cc(cc1[N+]([O-])=O)C#N

InChI

1S/C7H3IN2O3/c8-5-1-4(3-9)2-6(7(5)11)10(12)13/h1-2,11H

InChI key

SGKGVABHDAQAJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitroxinil is an anthelmintic agent, which is also an antiparasite. It is highly effective against Fasciola species and its mode of action involves uncoupling the oxidative phosphorylation in liver flukes and also reduces spermatogenesis in surviving flukes, resulting in fewer fertile eggs.

Application

Nitroxinil may be used as a reference standard in the determination of nitroxinil in veterinary active pharmaceutical ingredient manufacturing and bovine kidney using reversed-phase high-performance liquid chromatography (RP-HPLC) and liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Tarbin et al.
Journal of chromatography, 613(2), 347-353 (1993-04-02)
A high-performance liquid chromatographic method for the determination of the anthelmintic nitroxynil has been developed. The drug was extracted from cattle muscle tissue with 1% triethylamine in acetonitrile. The extract was evaporated to dryness and taken up in 0.1 M
L Moreno et al.
Research in veterinary science, 88(2), 315-320 (2009-11-07)
The effect of age/body weight in the plasma disposition kinetics of ivermectin (IVM) and nitroxynil (NTX) after their co-administration as a combined formulation to sheep was studied. Sixteen (16) male sheep were allocated into two experimental groups (n=8 each): (a)
Efficacy of nitroxynil against ivermectin, benzimidazole and salicylanilide resistant H contortus.
P C Jeannin et al.
The Veterinary record, 126(25), 624-625 (1990-06-23)
G Baziard-Mouysset et al.
Annales pharmaceutiques francaises, 54(5), 223-227 (1996-01-01)
Seven esters derivatives of Nitroxinil were prepared and their structures were assigned by IR and 1H-NMR spectroscopy. The rate of plasma and hepatic hydrolysis were evaluated in vitro in sheep and rabbit. In view of this profile of activity, pivaloyl
W J Blanchflower et al.
The Analyst, 114(9), 1013-1015 (1989-09-01)
A method is described for the determination of nitroxynil residues in muscle, liver and kidney. The samples were extracted into diethyl ether and cleaned-up using a simple liquid-liquid extraction step. Any nitroxynil present was separated from interfering compounds by high-performance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service