MABT833
Anti-Galectin-9 Antibody, clone 1G3
clone 1G3, from mouse
Synonym(s):
Galectin-9, Gal-9, Ecalectin, Tumor antigen HOM-HD-21, Galectin-9
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
1G3, monoclonal
species reactivity
human
technique(s)
ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... LGALS9(3965)
General description
Specificity
Immunogen
Application
Immunohistochemistry Analysis: A representative lot detected galectin-9 immunoreactivity in malignant cells using various paraffin-embedded nasopharangeal carcinoma (NPC) tissue sections (Barjon, C., et al. (2012). Infect Agent Cancer. 7(1):16-26).
Western Blotting Analysis: A representative lot detected endogenous galectin-9 isoforms in HeLa, lymphoblastoid cell line (LCL) REMB1, and nasopharyngeal carcinoma (NPC) cell lines C15 & C666-1, as well as exogenously expressed galectin-9 isoforms in transfected HeLa cells, but not in Burkitt’s lymphoma (BL) BL2 cells (Barjon, C., et al. (2012). Infect Agent Cancer. 7(1):16-26).
ELISA Analysis: A representative lot selectively captured human & murine galectin-9 (mGal-9 M, hGAL-9 S & M isoforms), but not human galectin 1, 2, 3, 4, 7, 8, or 10 (Barjon, C., et al. (2012). Infect Agent Cancer. 7(1):16-26).
Immunocytochemistry Analysis: A representative lot detected galectin-9 immunoreactivity in formalin-fixed and paraffin-embedded lymphoblastoid cell line (LCL) REMB1, but not in Burkitt’s lymphoma (BL) BL2 cells (Barjon, C., et al. (2012). Infect Agent Cancer. 7(1):16-26).
Quality
Western Blotting Analysis: 0.5 µg/mL of this antibody detected Galectin-9 in 10 µg of HeLa nuclear extract.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service