Skip to Content
Merck
All Photos(2)

Key Documents

MABT399

Sigma-Aldrich

Anti-CD36 Antibody, clone Long 6

clone Long 6, from mouse

Synonym(s):

Fatty acid translocase, FAT, Glycoprotein IIIb, GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, Platelet glycoprotein IV, GPIV, Thrombospondin receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
Long 6, monoclonal
application:
ICC
IP
WB
species reactivity:
human, rat
technique(s):
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
3

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Long 6, monoclonal

species reactivity

human, rat

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD36(948)

General description

CD36 is a major 88,000 MW platelet glycoprotein also found on monocytes, macrophages and some endothelial cells. It has multiple diverse roles in physiology. CD36 is a receptor for thrombospondin-1 (TSP-1), functioning as an inhibitor of angiogenesis. It also binds oxidized low density lipoproteins (LDL) and shed photoreceptor outer segments. CD36 is also act an endothelial-CAM acting in platelet adhesion and aggregration. Absence of CD36 protects against the formation of arterial plaques, thus slowing the development of atherosclerosis. CD36 transcription is regulated by the nuclear receptor PPAR-gamma.

Immunogen

GST-tagged recombinant protein corresponding to Human CD36.

Application

Research Category
Cell Structure
Research Sub Category
Immunological Signaling
This Anti-CD36 Antibody, clone Long 6 is validated for use in Western Blotting and Immunocytochemistry and Immunoprecipitation for the detection of CD36.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected CD36 in 10 µg of CD36 transfected MV3 cell lysate.
Western Blotting Analysis: A representative lot detected CD36 in CD36 transfected COS-7 cells (Thorne et al., 2006. DNA AND CELL BIOLOGY. Volume 25, Number 5).
Immunohistochemistry Analysis: A representative lot detected CD36 in GM-CSF elicited macrophages (Thorne et al., 2006. DNA AND CELL BIOLOGY. Volume 25, Number 5).
Immunoprecipitation Analysis: A representative lot immunoprecipitated CD36 in GM-CSF elicited macrophages (Thorne et al., 2006. DNA AND CELL BIOLOGY. Volume 25, Number 5).

Quality

Evaluated by Western Blotting in THP-1 cells treated with GM-CSF.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected CD36 in 10 µg GM-CSF treated THP-1 cell lysate.

Target description

~70 kDa observed. The calculated MW of this protein is 53 kDa It runs higher due to glycosolation. Uncharacterized bands may be observed in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joel James et al.
Arteriosclerosis, thrombosis, and vascular biology, 41(2), 734-754 (2020-12-11)
NFU1 is a mitochondrial iron-sulfur scaffold protein, involved in iron-sulfur assembly and transfer to complex II and LAS (lipoic acid synthase). Patients with the point mutation NFU1G208C and CRISPR/CAS9 (clustered regularly interspaced short palindromic repeats/clustered regularly interspaced short palindromic repeat-associated
Xuebing Wang et al.
Frontiers in physiology, 11, 1090-1090 (2020-09-29)
Hypoxic exercise may represent a novel therapeutic strategy to reduce and prevent obesity through the regulation of lipid metabolism. During hypoxic exercise, the targeting of peroxisome proliferator-activated receptor gamma (PPARγ) by miR-27b has been proposed to be one of the
Savanna N Weninger et al.
Microbiome, 11(1), 169-169 (2023-08-03)
Upper small intestinal dietary lipids activate a gut-brain axis regulating energy homeostasis. The prebiotic, oligofructose (OFS) improves body weight and adiposity during metabolic dysregulation but the exact mechanisms remain unknown. This study examines whether alterations to the small intestinal microbiota

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service