Skip to Content
Merck
All Photos(2)

Key Documents

MABF62

Sigma-Aldrich

Anti-Tim3 Antibody, clone 2E2

clone 2E2, from mouse

Synonym(s):

Hepatitis A virus cellular receptor 2 homolog, T-cell immunoglobulin and mucin domain-containing protein 3, T-cell membrane protein 3, TIMD-3, TIM-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E2, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HAVCR2(84868)

General description

As a member of the T cell immunoglobulin and mucin domain (TIM) family, Tim-3 is reported to promote or terminate Th1 immunity and is thought to influence a range of inflammatory conditions. It is constitutively expressed in a subset of murine IFNgamma-secreting Th1 cells, but not Th2 cells, in both human and mice. Additionally, Tim-3 is expressed dendritic cells, and by a subset of CD4+ cells where its levels of expression increase upon anti-CD3/28 stimulation. This molecule is also highly expressed in in vitro polarized Th1 cells, and at lower levels in Th17 cells. It is known to synergize with Toll-like receptors and is thought to regulate Th1 and Th17 cytokine secretion.

Immunogen

Recombinant protein corresponding to human Tim3.

Application

Anti-Tim3 Antibody, clone 2E2 detects level of Tim3 & has been published & validated for use in WB, FC.
Flow Cytometry Analysis: A 1:1,000 dilution of this antibody detected Tim3 in human monocyte derived dendritic cells.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blot in human thymus tissue lysate.

Western Blot Analysis: A 1:2,000 dilution of this antibody detected Tim3 in human thymus tissue lysate.

Target description

~30 kDa observed.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human thymus tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service