Skip to Content
Merck
All Photos(1)

Key Documents

MAB1924-C

Sigma-Aldrich

Anti-Laminin alpha5 Antibody, clone 4C7 (Ascites Free)

clone 4C7, from mouse

Synonym(s):

Laminin-10 subunit alpha, Laminin-11 subunit alpha, Laminin-15 subunit alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4C7, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
neutralization: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... LAMA5(3911)

General description

Laminin subunit alpha-5 (UniProt: O15230; also known as Laminin-10 subunit alpha, Laminin-11 subunit alpha, Laminin-15 subunit alpha) is encoded by the LAMA5 (also known as KIAA0533, KIAA1907) gene (Gene ID: 3911) in human. Laminins are a family of heterotrimeric extracellular glycoproteins consisting of alpha, beta, and gamma chains. Laminins mediate the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Thus far five alpha, three beta, and three gamma subunits are known, which assemble into at least 15 different laminin molecules. The alpha, beta, and gamma chains are bound to each other by disulfide bonds into a cross-shaped molecule comprising one long and three short arms with globules at each end. Alpha-5 is a subunit of laminin-10 (laminin-511), laminin-11 (laminin-521) and laminin-15 (laminin-523). It contains 22 laminin EGF-like domains, 5 5 laminin G-like domains, 1 laminin IV type A domain, and 1 laminin N-terminal domain. Laminin 5-alpha is expressed in heart, lung, kidney, muscle, pancreas, retina, and placenta. Very little or no expression is observed in brain and liver.

Specificity

Clone 4C7 reacts with the globular domain of the alpha5 chain of intact laminin and blocks the neurite stimulating activity of laminin.

Immunogen

Full length purified human laminin alpha 5.

Application

Anti-Laminin alpha5, clone 4C7 (Ascites Free), Cat. No. MAB1924-C, is a mouse monoclonal antibody that detects Laminin alpha 5 and has been tested for use in ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, and neutralization.
Immunohistochemistry Analysis: A representative lot detected Laminin alpha 5 in immunostained basement membranes in O.T.C.-embedded fetal and adult human lung cryosections (Pierce, R.A., et al. (2000). Am. J. Respir. Cell Mol. Biol. 23(6):742-747).

Neutralizing Analysis: A representative lot detected Laminin alpha 5 in adhesion of sickle cell patients (Hb SS) red blood cells (RBCs) to human placental laminin-coated surface (Lee, S.P., et al. (1998). Blood. 92(8):2951-2958).

ELISA Analysis: A representative lot detected Laminin alpha 5 in recombinant human (rh) Laminin-10 ( 5 1 1) and Laminin-11 ( 5 2 1), but not Laminin-8 ( 4 1 1) (Wondimu, Z., et al. (2013). PLoS One. 8(1):e53648).

Immunofluorescence Analysis: A representative lot detected Laminin alpha 5 in immunoreactivity in human skin and kidney cryosections (Wondimu, Z., et al. (2013). PLoS One. 8(1):e53648).

Immunoprecipitation Analysis: A representative lot immunoprecipitated Laminin alpha 5 in Laminin-10 ( 5 1 1) from A549 culture medium (Wondimu, Z., et al. (2013). PLoS One. 8(1):e53648).
Research Category
Cell Structure

Quality

Evaluated by Immunoprecipitation in A431 cell lysate.

Immunoprecipitation Analysis: 10 µg of this antibody immunoprecipitated Laminin alpha 5 in 500 µg of A431 cell lysate.

Target description

~260 kDa observed; 399.74 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing PBS

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service