KZ-1L-BKK
Intermediates for Further Manufacturing Use (FFMU) - RHCE (Rh) Specificities
Anti-E from the clone MS-16, Human IgM, Intermediate for further manufacturing use
Synonym(s):
Anti-e (clone MS-16), FFMU
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116102
NACRES:
NA.41
biological source
human
Quality Level
antibody form
unpurified
clone
MS-16, monoclonal
form
liquid
packaging
pack of 1 L
concentration
3-14 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
6.7-7.7
isotype
IgM
shipped in
ambient
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Standardized, concentrated cell culture supernatant, non-diafiltered, minimal pH buffering; For the formulation of Anti-e blood typing reagents or devices.Broad specificity, reactions are influenced by the presence or absence of the C antigen which makes it similar to an Anti-Ce; Contains 0.1% (w/v) Sodium Azide; Shelf Life: 24 months from date of filtration.Licensed by the US FDA under a shared manufacturing agreement
Application
Antibody intermediate for further manufacturing use.
Production of an in vitro blood typing product or device.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Partially processed reagent for further manufacture of non-injectable products only. The absence of all viral antigens has not been determined. No US Standard of Potency. Do not use if turbid. Contains 0.1%<nbsp/>(w/v) sodium azide.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service