Skip to Content
Merck
All Photos(1)

Key Documents

AG230

Sigma-Aldrich

Glial Fibrillary Acidic Protein, porcine

Synonym(s):

GFAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
eCl@ss:
32160405
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

Porcine

Quality Level

Assay

>99% (pure GFAP)

form

liquid

mol wt

calculated mol wt 49.44 kDa (428 a.a. spliced isoform; NP_001231326)
calculated mol wt 53.63 kDa (468 a.a. spliced isoform; XP_005668766)

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

ELISA: suitable
radioimmunoassay: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

General description

Glial fibrillary acidic protein (UniProt F1RR02) is encoded by the GFAP gene (Gene ID 396562) in porcine. GFAP is a type III intermediate filament (IF) thought to help maintain astrocyte mechanical strength and is closely related to vimentin, desmin, and peripherin, which are involved in the structure and function of cellular cytoskeleton. GFAP is expressed in astrocytes in the central nervous system (CNS), where it is involved in many important CNS processes, including cell communication and the blood brain barrier (BBC) function. GFAP contains the characteristic tripartite domain structure among IF members, a central α-helical rod domain (a.a. 73-377) flanked by a non-α-helical N-terminal head (a.a. 1-72) and a C-terminal tail (a.a. 378-432) domain. The head and tail domains are most structurally variable among IF proteins and their contributions to filament assembly vary depending on the IF proteins. For GFAP, the tail domain is reported to be important for filament assembly. The tail domain R416W mutation is linked to the neurological disorder Alexander disease (AxD).
Purified porcine Glial Fibrillary Acidic Protein (GFAP) purified from spinal cord.

Physical form

Liquid. Buffer = PBS.

Storage and Stability

Maintain at -20°C to -70°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

During shipment, small volumes of product will occasionally become entrapped in the seal of the product vial. For products with volumes of 200 μL or less, we recommend gently tapping the vial on a hard surface or briefly centrifuging the vial in a tabletop centrifuge to dislodge any liquid in the container′s cap.

Analysis Note

a 50 kDa single band by SDS-PAGE.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gallya Gannot et al.
The Journal of molecular diagnostics : JMD, 9(3), 297-304 (2007-06-27)
Layered peptide array is a new methodology for multiplex molecular measurements from two-dimensional life science platforms. The technology can be used in several different configurations depending on the needs of the investigator. Described here is an indirect layered peptide array
Michael Q Steinman et al.
General and comparative endocrinology, 193, 141-148 (2013-08-21)
Glia regulate the hypothalamic-pituitary-gonadal (HPG) axis in birds and mammals. This is accomplished mechanically by ensheathing gonadotrophin-releasing hormone I (GnRH) nerve terminals thereby blocking access to the pituitary blood supply, or chemically in a paracrine manner. Such regulation requires appropriate
Photostimulated expression of type 2 iodothyronine deiodinase mRNA is greatly attenuated in the rostral tuberal hypothalamus of the photorefractory turkey hen.
M Q Steinman,S C Dinius,T D Siopes,J R Millam
Journal of Neuroendocrinology null
Junchao Tong et al.
Neurobiology of disease, 82, 243-253 (2015-06-24)
Although gliosis is a normal response to brain injury, reports on the extent of astrogliosis in the degenerating substantia nigra in Parkinson's disease (PD) are conflicting. It has also been recently suggested that accumulation of nigral α-synuclein in this disorder
Sai-Sai Huang et al.
Molecular medicine reports, 22(6), 4821-4827 (2020-11-12)
Chronic postsurgical pain (CPSP) has a high incidence, but the underlying mechanism is not well understood. Accumulating evidence has suggested that central sensitization is the main mechanism of pain. To study the role of p120 in CPSP, a skin/muscle incision

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service