Skip to Content
Merck
All Photos(1)

Key Documents

8.41740

Sigma-Aldrich

Hexadecanedioic acid

for synthesis

Synonym(s):

Hexadecanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30O4
CAS Number:
Molecular Weight:
286.41
MDL number:
UNSPSC Code:
12352106
EC Index Number:
208-013-5
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥96.0% (acidimetric)

form

powder

mp

122-123 °C

storage temp.

2-30°C

SMILES string

OC(=O)CCCCCCCCCCCCCCC(=O)O

InChI

1S/C16H30O4/c17-15(18)13-11-9-7-5-3-1-2-4-6-8-10-12-14-16(19)20/h1-14H2,(H,17,18)(H,19,20)

InChI key

QQHJDPROMQRDLA-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 96.0 %
Melting range (lower value): ≥ 122 °C
Melting range (upper value): ≤ 126 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service