Skip to Content
Merck
All Photos(1)

Key Documents

8.20447

Sigma-Aldrich

2,3-Dichlorophenol

for synthesis

Synonym(s):

2,3-Dichlorophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2,3-(Cl)2C6H3OH
CAS Number:
Molecular Weight:
163.00
MDL number:
UNSPSC Code:
12352104
EC Index Number:
209-399-8
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

powder

bp

206 °C/1013 hPa

mp

57-60 °C

transition temp

flash point 115 °C

density

1.40 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

Clc1c(cccc1O)Cl

InChI

1S/C6H4Cl2O/c7-4-2-1-3-5(9)6(4)8/h1-3,9H

InChI key

UMPSXRYVXUPCOS-UHFFFAOYSA-N

Application

2,3-Dichlorophenol for synthesis. CAS 576-24-9, chemical formula 2,3-(Cl)₂C₆H₃OH.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 57 °C
Melting range (upper value): ≤ 60 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service