Skip to Content
Merck
All Photos(2)

Key Documents

8.16013

Sigma-Aldrich

Glycine

for synthesis

Synonym(s):

Gly, Aminoacetic acid, Aminoethanoic acid, Aminoacetic acid, Aminoethanoic acid, Glycocoll

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
635782
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-272-2
NACRES:
NA.22
grade:
for synthesis
technique(s):
inhibition assay: suitable
vapor pressure:
0.0000171 Pa ( 25 °C)

grade

for synthesis

Quality Level

vapor pressure

0.0000171 Pa ( 25 °C)

Assay

≥99.0% (perchloric acid titration)

form

solid

potency

7930 mg/kg LD50, oral (Rat)

technique(s)

inhibition assay: suitable

pH

5.9-6.4 (20 °C, 50 g/L in H2O)

pKa (25 °C)

(1) 2.35, (2) 9.60
2.35

mp

240 °C (dec.) (lit.)

density

1.161 g/cm3 at 20 °C

bulk density

920 kg/m3

storage temp.

2-30°C

SMILES string

NCC(O)=O

InChI

1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)

InChI key

DHMQDGOQFOQNFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Identity (IR): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service