Skip to Content
Merck
All Photos(1)

Key Documents

8.01647

Sigma-Aldrich

Boron trifluoride-diethyl ether complex

for synthesis

Synonym(s):

Boron trifluoride-diethyl ether complex, Boron trifluoride-ethyl etherate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10O · BF3
CAS Number:
Molecular Weight:
141.93
MDL number:
UNSPSC Code:
12352301
EC Index Number:
203-689-8
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

2.35 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

185 °C (DIN 51794)

expl. lim.

5.1-18.2 % (v/v)

kinematic viscosity

1.69 cSt(20 °C)

mp

-58 °C

transition temp

flash point 58.5 °C (DIN 51758)

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

B(F)(F)F.O(CC)CC

InChI

1S/C4H10O.BF3/c1-3-5-4-2;2-1(3)4/h3-4H2,1-2H3;

InChI key

KZMGYPLQYOPHEL-UHFFFAOYSA-N

Application

  • Synthesis of a new boron complex with imine ligand: Synthesis, characterization and fluorescent properties: This article discusses the synthesis and properties of a novel boron complex with imine ligand, highlighting its fluorescent characteristics (Şen, 2019).
  • Facile synthesis of diphenylmethyl esters from 2-diphenylmethoxypyridine using catalytic boron trifluoride· diethyl etherate: A method for synthesizing diphenylmethyl esters employing boron trifluoride· diethyl etherate as a catalyst is presented (La and Kim, 2018).
  • An impedance-morphology study on poly (3-methylthiophene) coated electrode obtained in boron trifluoride diethyl etherate–acetonitrile: This research examines the effects of boron trifluoride diethyl etherate on the impedance and morphology of polythiophene coatings (Dolas and Sarac, 2014).
  • A fast and practical synthesis of tert-butyl esters from 2-tert-butoxypyridine using boron trifluoride· diethyl etherate under mild conditions: The article details a synthesis technique for tert-butyl esters using boron trifluoride· diethyl etherate, emphasizing its efficiency and mild reaction conditions (La and Kim, 2018).

Analysis Note

Assay BF3 (acidimetric): 48.0 - 52.0 %
Density (d 20 °C/ 4 °C): 1.120 - 1.130

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT RE 1 Inhalation

Target Organs

Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

137.3 °F - closed cup

Flash Point(C)

58.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service