Skip to Content
Merck
All Photos(2)

Key Documents

73083BC

Millipore

GDS Gel Cooling Block

BioControl, For use with GDS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103037
Pricing and availability is not currently available.

manufacturer/tradename

BioControl Systems 73083

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

room temp

Components

One box of 2 cooling blocks

Testing Method

PCR

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service