Skip to Content
Merck
All Photos(1)

Key Documents

73004BC

Millipore

GDS Adhesive Sheets

BioControl, For use with GDS

Synonym(s):

Sealing film

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121814

description

For use with PICKPEN™ PIPETMAX™ system

form

sheet

manufacturer/tradename

BioControl Systems 73004

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

storage temp.

room temp

General description

GDS Adhesive Sheets are the consumable accessories of the Assurance® GDS assays. Assurance® GDS is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples. The sealing film strip measures 79.4 x 139.7 mm and offers sufficient sealing area for all PCR plates. The length between the perforations with end tabs removed is 125.1 mm. These adhesive sheets are a reliable method of sealing plates for PCR applications. The high integrity sealing materials give the best protection against evaporation for high efficiency PCR.

Application

The Assurance® GDS Adhesive Sheets are sealing film to provide effective and airtight sealing and help protect PCR samples during sample preparation.

Features and Benefits

  • Strong adhesive prevents evaporation during thermal cycling or sample storage
  • No sticky residue on the plate after the sheet is peeled off
  • End tabs for easy seal application and removal
  • Clear for sample visualization and inspection
  • Temperature range: -20°C to 120°C

Components

Box of 100 film sheets

Compatibility

BioControl GDS

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service