Skip to Content
Merck
All Photos(1)

Key Documents

681641

Sigma-Aldrich

Wee1 Inhibitor II

The Wee1 Inhibitor II, also referenced under CAS 622855-50-9, controls the biological activity of Wee1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Wee1 Inhibitor II, 6-Butyl-4-(2-chlorophenyl)-9-hydroxypyrrolo[3,4-c]carbazole-1,3-(2H,6H)-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H19ClN2O3
CAS Number:
Molecular Weight:
418.87
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 10 mg/mL
ethanol: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A pyrrolocarbazole compound that acts as a potent, ATP-binding site-targeting inhibitor of Wee1 (IC50 = 59 nM) with a ~590-fold selectivity over the related checkpoint kinase Chk1 (IC50 = 35 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Wee1
Product does not compete with ATP.
Reversible: no
Target IC50: 59 nM against Wee1

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Palmer, B.D., et al. 2006. J. Med. Chem.49, 4896.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service