Skip to Content
Merck
All Photos(1)

Key Documents

372040

Sigma-Aldrich

Haspin Kinase Inhibitor, CHR-6494

Synonym(s):

Haspin Kinase Inhibitor, CHR-6494, Indazolyl-propylimidazopyridazin-amine, 3-(1H-Indazol-5-yl)-N-propylimidazo[1,2-b]pyridazin-6-amine, Haploid Germ Cell-Specific Nuclear Protein Kinase Inhibitor, CHR-6494

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N6
CAS Number:
Molecular Weight:
292.34
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.54
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[n]21nc(ccc2ncc1c3cc4c([nH]nc4)cc3)NCCC

InChI

1S/C16H16N6/c1-2-7-17-15-5-6-16-18-10-14(22(16)21-15)11-3-4-13-12(8-11)9-19-20-13/h3-6,8-10H,2,7H2,1H3,(H,17,21)(H,19,20)

InChI key

CZZCAOGIEGXMBZ-UHFFFAOYSA-N

General description

A cell-permeable indazolylimidazopyridazinamine compound that acts as a potent and reversible inhibitor of histone kinase haspin activity (IC50 = 2 nM) with moderate selectivity over TrkA, GSK-3β, PIM1, Cdk1/B and Cdk2/A (% inhibition at 100 nM = 58, 48, 36, 34 and 33, respectively) among a panel of 27-kinases. Shown to block pH3-Thr3, with no effect on pH3-Ser10 and pH3-Ser28, cause mitotic catastrophe, upregulate BUB1 and cyclin B1 levels, and potently induce apoptosis (IC50 = 473, 500 and 752 nM in HeLa, HCT-116 and MDA-MB-231 cells, respectively). Further, efficiently blocks bFGF-induced sprouting vessel by 70% at 1 µM (chicken embryo aortic arch ring assay) and suppresses tumor growth in HCT-116 xenografted mouse model (50 mg/kg, i.p.).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliqout and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Huertas, D., et al. 2012. Oncogene31, 1408.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hisayo Nishida-Fukuda et al.
PloS one, 16(4), e0249912-e0249912 (2021-04-15)
HASPIN is a serine/threonine kinase that regulates mitosis by phosphorylating histone H3 at threonine 3. The expression levels of HASPIN in various cancers are associated with tumor malignancy and poor survival, suggesting that HASPIN inhibition may suppress cancer growth. As

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service