188079
Anolyte for coulometric Karl Fischer Titration without diaphragm Aquastar®
Synonym(s):
Karl Fischer Anolyte Solution
About This Item
Recommended Products
material
diaphragm (No)
reaction suitability
reaction type: coulometric
technique(s)
Karl-Fischer titration: suitable
titration: suitable
transition temp
flash point 12.5 °C
suitability
passes test for application
storage temp.
15-25°C
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
54.5 °F
Flash Point(C)
12.5 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
New Aquastar® Karl Fischer Anolyte for precise and reproducible determination of low water content in the range of 10 ppm to 10000 ppm for cells without a diaphragm.
Methods including internal extraction, external extraction, and Karl Fischer oven ensure accurate water content determination by titration.
Protocols
Karl Fischer titration offers accurate water content determination in hemp samples compared to the loss on drying method.
Karl Fischer titration offers accurate water content determination in hemp samples compared to the loss on drying method.
Karl Fischer titration offers accurate water content determination in hemp samples compared to the loss on drying method.
Karl Fischer titration offers accurate water content determination in hemp samples compared to the loss on drying method.
Related Content
Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.
Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.
Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.
Complete offer for petrochemical testing including environmental and industrial hygiene monitoring solutions.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service