Skip to Content
Merck
All Photos(1)

Key Documents

1.51911

Supelco

Purospher®STAR Si (5 µm) HPLC Columns

L × I.D. 250 mm x 4.6 mm Hibar® RT HPLC Column

Synonym(s):

Purospher® STAR SI (5µm) Hibar® RT 250-4.6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52
column L × I.D.:
25 cm × 4.6 mm
surface area:
330 m2/g
particle size:
5 μm
matrix active group:
Silica bonding phase
pore size:
120 Å pore size
matrix:
fully porous particle

Product Name

Purospher® STAR Si (5 µm), L × I.D. 250 mm x 4.6 mm Hibar® RT HPLC column

material

stainless steel column

Agency

suitable for USP L3

product line

Purospher®

feature

endcapped: no

packaging

1 ea of

parameter

400 bar pressure
65 °C max. temp.

technique(s)

HPLC: suitable
LC/MS: suitable
mass spectrometry (MS): suitable (High)

column L × I.D.

25 cm × 4.6 mm

surface area

330 m2/g

matrix

fully porous particle

matrix active group

Silica bonding phase

particle size

5 μm

pore size

120 Å pore size

operating pH range

1.5-10.5

separation technique

hydrophilic interaction (HILIC)
normal phase
supercritical fluid (SFC)

storage temp.

no temp limit

Looking for similar products? Visit Product Comparison Guide

General description

Purospher® STAR Si endcapped, 5 µm HPLC columns offers highest separation efficiency for normal-phase chromatography of low molecular weight compounds soluble in organic solvents.Purospher® STAR Si, 5 µm Hibar® RT 250-4.6 is a ready to use stainless steel column suitable for connection to any HPLC system.The recommended guard column is Purospher® STAR Si (5 µm) LiChroCART 4-4 part number 1.50249.0001. For direct connection to a Hibar® RT columns the Precolumn holder for 4-4 LiChroCART cartridges, part number 1.51487.0001 can be used.The column dimension of 250 mm length x 4.6 mm I.D. is the optimal choice for the HPLC separation of complex samples allowing significant solvent savings and improved sensitivity.Learn more about Purospher® STAR HPLC and UHPLC ColumnsLearn more about the Supelco HPLC Column portfolio and Method development:Brochure: HPLC and UHPLC Column Selection GuideBrochure: A Practical Guide to HPLC - Method Development Guide

Application

Intended to be used in the separation of aflatoxins, carotenoids, oils, fat soluble vitamins, lipids, and fatty acids.

Features and Benefits

  • Ultra-pure silica (99.999%) for excellent peak symmetry
  • High separation efficiency
  • Reproducible results from run-to-run and batch-to-batch
  • Use with Normal-phase or HILIC conditions (needs to be flushed to HILIC mobile phase)
  • Good suitability for LC-MS applications

Legal Information

Hibar is a registered trademark of Merck KGaA, Darmstadt, Germany
LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service