Skip to Content
Merck
All Photos(2)

Documents

1.46569

Millipore

NaCl Solution (phys.)

bottle of, ready-to-use, bottle volume 100 mL , filling volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type screw cap with 3 septa
ready-to-use

packaging

bottle of

technique(s)

direct inoculation: suitable
sterile filtration: suitable

bottle capacity

125 mL

bottle volume

100 mL , filling volume

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

NaCl Solution (phys.) is an isotonic buffer to maintain the osmotic balance in the test suspension. The unique formula acts as a general diluent which maintains viability of test microorganisms present in sample.
The solution does not promote microbial growth due to the absence of nutrients.

Application

NaCl Solution (phys.) is used as an isotonic diluent in various area. It is also recommended for preparing test suspensions of microorganisms.

Features and Benefits

  • Our ready-to-use media provide the highest level of quality and testing confidence. They have been formulated and tested to meet the pharmacopoeia requirements.
  • Sterility testing media and rinse solutions are manufactured in an ISO 9001, environmentally controlled production center. Each lot undergoes a stringent quality control (QC) procedure, including pH, sterility, and growth promotion tests.
  • Our manufacturing approach ensures the highest level of clarity for our media and rinsing fluids, therefore improving accuracy, and significantly reducing the risk of incorrect interpretation and false results.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service