1.15265
Heating bath fluid
for heating baths up to approx. 170°C
Synonym(s):
Heating bath fluid
About This Item
Recommended Products
Quality Level
form
liquid
autoignition temp.
390 °C
potency
>2000 mg/kg LD50, oral (Rat)
pH
6-8 (20 °C, 100 g/L in H2O)
viscosity
~330 mPa.s(20 °C)
kinematic viscosity
220-280 cSt(23 °C)
transition temp
flash point >290 °C
density
1.15 g/cm3 at 23 °C
storage temp.
no temp limit
SMILES string
O(C(COCCO)COCCO)CCO
InChI
1S/C9H20O6/c10-1-4-13-7-9(15-6-3-12)8-14-5-2-11/h9-12H,1-8H2
InChI key
UCYLROVJSUACAD-UHFFFAOYSA-N
Application
Analysis Note
Suitability for heating bath liquid: passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service