Skip to Content
Merck
All Photos(1)

Key Documents

1.07225

Supelco

1,10-Phenanthroline monohydrate

GR ACS

Synonym(s):

1,10-Phenanthroline monohydrate, o-Phenanthroline monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2 · xH2O
CAS Number:
Molecular Weight:
180.21 (anhydrous basis)
MDL number:
UNSPSC Code:
12352005
EC Index Number:
200-629-2
NACRES:
NA.21
form:
solid
Pricing and availability is not currently available.

Quality Level

form

solid

potency

132 mg/kg LD50, oral (Rat)

mp

93-94 °C

solubility

3.3 g/L

bulk density

300 kg/m3

storage temp.

15-25°C

SMILES string

n1c2c3ncccc3ccc2ccc1.O

InChI

1S/C12H8N2.H2O/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;/h1-8H;1H2

InChI key

PPQJCISYYXZCAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Linkage

Replaces: PX0460-5; PX0460

Analysis Note

Assay (Perchloric acid titration, calc. on anhydrous substance): ≥ 99.5 %
Identity (IR-spectrum): conforms
Suitability as redox indicator: conforms
Suitability for determining iron: conforms
Melting point (anhydrous substance): 117 - 120 °C
substances insoluble in ethanol: conforms
Water (according to Karl Fischer): 8.5 - 11.0 %

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service