Skip to Content
Merck
All Photos(2)

Documents

1.06580

Supelco

di-Sodium hydrogen phosphate dihydrate

for analysis EMSURE®

Synonym(s):

di-Sodium hydrogen phosphate dihydrate, Sodium monohydrogen phosphate, Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, Sodium phosphate dibasic dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 2H2O
CAS Number:
Molecular Weight:
177.99
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-448-7
NACRES:
NA.21

grade

Laboratory Reagent

Quality Level

product line

EMSURE®

Assay

≥99.5% (acidimetric)

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

pH

9.0-9.2 (20 °C, 50 g/L in H2O)

mp

92.5 °C (Elimination of water of crystallisation)

solubility

93 g/L

density

2.1 g/cm3 at 20 °C

bulk density

850‑1000 kg/m3

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Cu: ≤0.0003%
Fe: ≤0.001%
K: ≤0.005%
N: ≤0.001%
Pb: ≤0.001%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

SMILES string

O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.2H2O/c;;1-5(2,3)4;;/h;;(H3,1,2,3,4);2*1H2/q2*+1;;;/p-2

InChI key

KDQPSPMLNJTZAL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Linkage

Replaces: 1.06580.9057; SX0713; SX0713-5

Analysis Note

Assay (acidimetric): ≥ 99.5 %
Identity: passes test
pH-value (5 %; water): 9.0 - 9.2
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
Cu (Copper): ≤ 0.0003 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.005 %
Pb (Lead): ≤ 0.001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Application Note: Peroxidase in vegetables – Visual determination after conversion of a specific substrate.

Application Note: Peroxidase in vegetables – Visual determination after conversion of a specific substrate.

Application Note: Peroxidase in vegetables – Visual determination after conversion of a specific substrate.

Application Note: Peroxidase in vegetables – Visual determination after conversion of a specific substrate.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service