Skip to Content
Merck
All Photos(1)

Key Documents

103882

SAFC

Cell Culture Media 4CHO Sample Kit

powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104105
Pricing and availability is not currently available.

Quality Level

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

shipped in

ambient

storage temp.

2-8°C

General description

The Cell Culture Media 4CHO Sample Kit contains 1L equivalent of the following products:
  • 24370C EX-CELL® Advanced HD Perfusion Medium
  • 103840 Cellvento® 4CHO-X Expansion Medium
  • 24366C EX-CELL® Advanced CHO Fed-batch Medium
  • 103795 Cellvento® 4CHO Medium
  • 103796 Cellvento® 4Feed
  • 24368C EX-CELL® Advanced Feed 1

Application

Cell culture media sample kit to support media evaluation for fed-batch and perfusion applications and process intensification.

Features and Benefits

The products support evaluation for achieving high cell densities in cell expansion phase (seed train), identifing the optimal combination of basal medium and feed in fed-batch and a designed medium for perfusion processing.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • 24370CEX-CELL® Advanced HD Perfusion Medium, powderSDS

  • 1.03840Cellvento® 4CHO-X COMP, expansion mediumSDS

  • 24366CEX-CELL® Advanced CHO Fed-batch Medium, powderSDS

  • 1.03795Cellvento® 4CHO COMP, chemically defined cell culture medium, ≤1.0 IU/mL endotoxin, pH 5.1-5.6(without supplement), pH 6.7-7.3(with supplement)SDS

  • 1.03796Cellvento® 4Feed COMP, chemically defined cell culture feedSDS

  • 24368CEX-CELL® Advanced CHO Feed 1 (without glucose)SDS

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore how low-impurity iron sources enhance consistency and control in recombinant protein production using Cellvento® 4CHO medium.

Explore how low-impurity iron sources enhance consistency and control in recombinant protein production using Cellvento® 4CHO medium.

Explore how low-impurity iron sources enhance consistency and control in recombinant protein production using Cellvento® 4CHO medium.

Explore how low-impurity iron sources enhance consistency and control in recombinant protein production using Cellvento® 4CHO medium.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service