Skip to Content
Merck
All Photos(2)

Key Documents

1.00863

Supelco

Ethyl acetate

for spectroscopy Uvasol®

Synonym(s):

Ethyl ethanoate, Acetic ester, Ethyl ester, EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
12191502
EC Index Number:
205-500-4
NACRES:
NA.03
technique(s):
UV/Vis spectroscopy: suitable
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3 (20 °C, vs air)

Quality Level

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

801 °F

potency

5620 mg/kg LD50, oral (Rat)
>18000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.01% Water

evapn. residue

≤0.0002%

color

APHA: ≤10

transmittance

255 nm, ≥20%
260 nm, ≥75%
263 nm, ≥80%
265 nm, ≥90%
270 nm, ≥98%

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

transition temp

flash point -4 °C

solubility

85.3 g/L

density

0.902 g/mL at 25 °C (lit.)

UV absorption

λ: 255 nm Amax: ≤0.70
λ: 260 nm Amax: ≤0.13
λ: 263 nm Amax: ≤0.10
λ: 265 nm Amax: ≤0.05
λ: 270 nm Amax: ≤0.01

format

neat

storage temp.

2-30°C

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol<TMSYMBOL></TMSYMBOL> solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol<TMSYMBOL></TMSYMBOL> solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.

Application

  • Investigation of anti-depression effects and potential mechanisms of the ethyl acetate extract of Cynomorium songaricum Rupr. through the integration of in vivo experiments, LC-MS/MS chemical analysis, and a systems biology approach.: This article describes the application of ethyl acetate in the extraction of antidepressant compounds, integrating chemical analysis with biological testing to explore therapeutic potentials (Zhang et al., 2023).
  • Steroid Components of Marine-Derived Fungal Strain Penicillium levitum N33.2 and Their Biological Activities.: The use of ethyl acetate for extracting steroid compounds from marine fungi is discussed, focusing on the biological activities and potential pharmaceutical applications of these extracts (Hoang et al., 2023).
  • Streptomyces sp. MNP32, a forest-dwelling Actinomycetia endowed with potent antibacterial metabolites.: This study details the extraction of antibacterial compounds using ethyl acetate from a forest-dwelling Actinomycetes species, contributing to the discovery of new antibiotics (Konwar et al., 2023).

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

UVASOL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F

Flash Point(C)

-4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service