Skip to Content
Merck
All Photos(4)

Key Documents

06-659

Sigma-Aldrich

Anti-phospho-c-Jun (Ser73) Antibody

Upstate®, from rabbit

Synonym(s):

Activator protein 1, Jun activation domain binding protein, Proto-oncogene c-jun, V-jun avian sarcoma virus 17 oncogene homolog, enhancer-binding protein AP1, jun oncogene, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

Xenopus (based on 100% sequence homology), chicken (based on 100% sequence homology), rat (based on 100% sequence homology), bovine (based on 100% sequence homology), zebrafish (based on 100% sequence homology)

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer73)

Gene Information

human ... JUN(3725)
mouse ... Jun(16476)
rat ... Jun(24516)
zebrafish ... Jun(335916)

General description

c-Jun is a component of the transcription factor AP-1 that binds and activates transcription at TRE/AP-1 elements and appears to be a major downstream target of the SAPK/JNK signaling pathway. The transcriptional activity of c-Jun is regulated by phosphorylation at Ser63 and Ser73. Extracellular signals including growth factors, transforming oncoproteins and UV irradiation stimulate phosphorylation of c-Jun at Ser63/73 and activate c-Jun dependent transcription. Mutation of Ser63/73 renders c-Jun nonresponsive to mitogenic and stress induced

signaling pathways. The MAP kinase homologue, SAPK/JNK, binds to the N-terminal region of c-Jun and phosphorylates c-Jun at Ser63/73. In addition, the activity of SAPK/JNK is stimulated by the same signals that activate c-Jun.

Specificity

This antibody detects phospho-c-Jun. May also detect phospho-JunD in properly treated cells since the sequence homology is identical (13/13).

Immunogen

KLH-conjugated linear peptide corresponding to the human Phospho-C-Jun (Ser73) near the N-terminus.

Application

Anti-phospho-c-Jun (Ser73) Antibody is a Rabbit Polyclonal Antibody for detection of phospho-c-Jun (Ser73) also known as Activator protein 1 & has been validated in ICC, IP & WB.
Western Blot Analysis :
A previous lot of this antibody detected 10 ng of phosphorylated gst-Jun, but did not detect 10 ng of nonphosphorylated Jun, as reported by an independent laboratory.

Quality

Evaluated by Western Blotting in Anisomycin treated NIH/3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Phospho-C-Jun (Ser73) in 10 µg of Anisomycin treated NIH/3T3 cell lysate.

Target description

~40 kDa

Linkage

Replaces: 04-213

Physical form

Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH7.4), 150 mM NaCl with 0.05% sodium azide.

Analysis Note

Control
Untreated and anisomycin-treated NIH/3T3 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos O Stocco et al.
The Journal of biological chemistry, 277(5), 3293-3302 (2001-11-24)
We have previously demonstrated that prostaglandin F(2alpha) (PGF(2alpha)) induces a rapid and transient expression of Nur77 in luteal cells. We have shown that Nur77 plays an important role in ovarian physiology by mediating the PGF(2alpha) induction of 20alpha-HSD, a steroidogenic
C Urbich et al.
The Journal of clinical investigation, 108(10), 1451-1458 (2001-11-21)
Atherosclerosis is an inflammatory disease of large arteries that is initiated through the activation of endothelium by proinflammatory mediators. CD40 receptor stimulation has been implicated in the pathogenesis of atherosclerosis. One of the most important atheroprotective stimuli is the viscous
Daniel Spohn et al.
Molecular pharmacology, 78(5), 865-876 (2010-08-18)
Thapsigargin is a specific inhibitor of the sarco/endoplasmic reticulum Ca(2+) ATPase of the endoplasmic reticulum. Here, we show that stimulation of human HaCaT keratinocytes with nanomolar concentrations of thapsigargin triggers expression of activating transcription factor (ATF) 3, a basic-region leucin
M Repici et al.
Neuroscience, 159(1), 94-103 (2009-01-13)
The c-Jun N-terminal kinase (JNK) is a mitogen-activated protein kinase (MAPK) activated by stress-signals and involved in many different diseases. Previous results proved the powerful effect of the cell permeable peptide inhibitor d-JNKI1 (d-retro-inverso form of c-Jun N-terminal kinase-inhibitor) against
Stable expression of intracellular Notch suppresses v-Src-induced transformation in avian neural cells.
Mateos, S; Amarir, S; Laugier, D; Marx, M; Calothy, G
Oncogene null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service