Skip to Content
Merck
All Photos(1)

Key Documents

L-023

Supelco

Levetiracetam-D6 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Deuterated Levetiracetam Solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D6H8N2O2
Molecular Weight:
176.25
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])(N1CCCC1=O)C(N)=O

InChI

1S/C8H14N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h6H,2-5H2,1H3,(H2,9,12)/i1D3,2D2

InChI key

HPHUVLMMVZITSG-ZBJDZAJPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Suitable uses for this certified solution standard include as an internal standard for quantitation of levetiracetam levels in biological matrices such as urine or isotope dilution methods by LC/MS or GC/MS for clinical toxicology, urine drug testing, or forensic analysis applications. Levetiracetam, marketed under the trade name Keppra®, is an antiepileptic drug used primarily for treatment of seizures along with occasional use in treating neuropathic pain.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Keppra is a registered trademark of UCB Pharma, S.A.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service