Skip to Content
Merck
All Photos(1)

Key Documents

I-056

Supelco

Isotonitazene-13C6 hydrochloride solution

100 μg/mL in methanol ((as free base)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C1713C6H30N4O3 · HCl
Molecular Weight:
452.93
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol ((as free base))

format

single component solution

storage temp.

−20°C

SMILES string

CC(C)OC(C=C1)=CC=C1CC2=N[13C]3=[13C](N2CCN(CC)CC)[13CH]=[13CH][13C]([N+]([O-])=O)=[13CH]3.Cl

InChI

1S/C23H30N4O3.ClH/c1-5-25(6-2)13-14-26-22-12-9-19(27(28)29)16-21(22)24-23(26)15-18-7-10-20(11-8-18)30-17(3)4;/h7-12,16-17H,5-6,13-15H2,1-4H3;1H/i9+1,12+1,16+1,19+1,21+1,22+1;

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service