Skip to Content
Merck
All Photos(1)

Key Documents

900415C

Avanti

Oleic Acid (18-azido)

Avanti Research - A Croda Brand, chloroform solution

Synonym(s):

(Z)-18-azidooctadec-9-enoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H33N3O2
CAS Number:
Molecular Weight:
323.47
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 25 mg (900415C-25mg)
pkg of 1 × 5 mg (900415C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL in chloroform (900415C-25mg)
5 mg/mL in chloroform (900415C-5mg)

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

Packaging

pkg of 1 x 25 mg, 10 mg/mL, 2.5 mL; 5mL Clear Glass Sealed Ampule (900415C-25mg)
pkg of 1 x 5mg, 5 mg/mL, 1 mL; 5mL Clear Glass Sealed Ampule (900415C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre-Christian Violet et al.
JCI insight, 5(1) (2019-12-11)
BACKGROUNDWe hypothesized that obesity-associated hepatosteatosis is a pathophysiological chemical depot for fat-soluble vitamins and altered normal physiology. Using α-tocopherol (vitamin E) as a model vitamin, pharmacokinetics and kinetics principles were used to determine whether excess liver fat sequestered α-tocopherol in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service