Skip to Content
Merck
All Photos(2)

Key Documents

852448C

Avanti

18:0 dimethylacetal

octadecanal-dimethylacetal, chloroform

Synonym(s):

1,1-Dimethoxyoctadecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H42O2
CAS Number:
Molecular Weight:
314.55
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (852448C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 852448C

concentration

10 mg/mL (852448C-10mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(OC)OC

InChI key

YDFMLZBWAXMUEX-UHFFFAOYSA-N

General description

DMAs are an indirect marker of levels of plasmalogens in plasma and may be used as standards for quantitation of plasmalogen in biological samples.

Application

18:0 dimethylacetal has been used to compare placental plasmalogen fatty acid profiles of normotensive and preeclampsia (PE) pregnancies.[1]

Packaging

5 mL Clear Glass Sealed Ampule (852448C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Brien et al.
Placenta, 51, 82-88 (2017-03-16)
Preeclampsia (PE) was shown to affect the placental content and the transfer of polyunsaturated fatty acids (PUFA) to the fetus. Plasmalogens, a type of phospholipids with a vinyl-ether link at the sn-1 position, play an antioxidant role and are specifically
Marina Moraitou et al.
Blood cells, molecules & diseases, 41(2), 196-199 (2008-05-27)
Plasmalogens represent a unique type of phospholipids characterized by the presence of a vinyl-ether bond at the sn-1 position of the glycerol backbone. Peroxisomes are essential in their biosynthesis. Their suggested functions include protection against oxidative stress, participation in signal
T Brosche
Archives of gerontology and geriatrics, 32(3), 283-294 (2001-06-08)
The precise role played by plasmalogen phospholipids (PL) of lipoproteins and cell membranes is not well understood. However, they might act as endogenous antioxidants in defending cell membranes and lipoproteins from reactive oxygen species. A decline of plasmalogen concentrations has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service