Skip to Content
Merck
All Photos(1)

Key Documents

791282P

Avanti

α-mycolic acid, keto cis

powder, Avanti Research - A Croda Brand 791282P

Synonym(s):

α-mycolic acid, keto cis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C86H168O4
CAS Number:
Molecular Weight:
1266.25
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (791282P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791282P

lipid type

bacterial lipids

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Mycolic acid is a long Fatty acid found in the cell walls of the mycolata taxon, a group of bacteria that includes Mycobacterium Tuberculosis, the causative agent of the disease tuberculosis.
Mycolic acid is composed of 70-90 carbons, with the saturated α-chain made up of 24 carbons and meromycolate chains made up of 40-60 carbons. Nature or structure of mycolic acid facilitates the determination of fluidity and permeability of mycobacterial cell wall.

Packaging

5 mL Amber Glass Screw Cap Vial (791282P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mycolic acid structure determines the fluidity of the mycobacterial cell wall
Liu J, et al.
Test, 271(47), 29545-29551 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service