Skip to Content
Merck
All Photos(1)

Key Documents

330736W

Avanti

Bile Acid SPLASH

Avanti Research - A Croda Brand

Synonym(s):

Bile Acid SPLASH LIPIDOMIX Quantitative Mass Spec Internal Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.21

form

methanol solution

packaging

pkg of 1 x 1 ea (330736W-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

technique(s)

mass spectrometry (MS): suitable

shipped in

dry ice

storage temp.

−20°C

Features and Benefits

  • Save time: pre-mixed internal standards.
  • Save money: 1 vial is sufficient for 100 plasma samples.
  • Exact concentrations provided for quantitative lipidomic analysis.
  • Applicable to multiple MS approaches.

Packaging

2 mL Amber Glass Sealed Ampule (330736W-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
SPLASH is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 330256WCholic acid-d4, Avanti Research - A Croda Brand 1150nMSDS

  • 330278WTaurocholic acid-d4 sodium salt, Avanti Research - A Croda Brand 1000nMSDS

  • 330277WGlycocholic acid-d4, Avanti Research - A Croda Brand 1150nMSDS

  • 330257WDeoxycholic acid-d4, Avanti Research - A Croda Brand 1650nMSDS

  • 330272WTaurodeoxycholic acid-d4 sodium salt, Avanti Research - A Croda Brand 1000nMSDS

  • 330271WGlycodeoxycholic acid-d4, Avanti Research - A Croda Brand 1275nMSDS

  • 330259WChenodeoxycholic acid-(2,2,4,4-d4), Avanti Research - A Croda Brand 1150nMSDS

  • 330274WTaurochenodeoxycholic acid-D4 sodium salt, Avanti Research - A Croda Brand 1000nMSDS

  • 330273WGlycochenodeoxycholic acid-d4, Avanti Research - A Croda Brand 3800nMSDS

  • 330279WTauroursodeoxycholic acid-d4, sodium salt, Avanti Research - A Croda Brand 500nMSDS

  • 330276WGlycoursodeoxycholic acid-D4, Avanti Research - A Croda Brand 600nMSDS

  • 330839W3β,5α,6β-trihydroxycholanoic acid-d4, Avanti Research - A Croda Brand 250nMSDS

  • 330258Wlithocholic acid-d4, Avanti Research - A Croda Brand 250nMSDS

  • 330280WTaurolithocholic acid-d4 sodium salt, Avanti Research - A Croda Brand 250nMSDS

  • 330275WGlycolithocholic acid-D4, Avanti Research - A Croda Brand 250nMSDS

  • 330260WUrsodeoxycholic acid-d4, Avanti Research - A Croda Brand 300nMSDS

  • 330840WCholenic acid-d4, Avanti Research - A Croda Brand 250nMSDS

See All (17)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service