Key Documents
Z566756
Scienceware® Secador® desiccator cabinet
model 1.0, horizontal profile, amber
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Pricing and availability is not currently available.
material
polycarbonate
feature
Can be stacked three high.
packaging
pack of 1 ea
manufacturer/tradename
model 1.0
Bel-Art F42071-0008
H × W × D
21.3 cm × 34.1 cm × 41.4 cm
door opening H × W
11.4 cm × 22.5 cm
color
amber
General description
Dry storage of humidity sensitive products is now easier. Secador cabinets assure moisture and dust free storage of valuable items like reagents and pipettes with their proprietary sealed design.
Available in small or large models, with either horizontal or vertical orientation, these uniquely-designed units are available totally clear, clear with blue end caps, and amber. The stackable small model is perfect for maximizing storage space in a minimum footprint.
Cabinets are molded of polycarbonate resistant to staining, crazing and chemical attack and have built-in protection that blocks 99% of UV light. Desiccants can be placed under the bottom shelf or on any shelf. A hygrometer in the door allows for easy monitoring of relative humidity. Carrying handles make them easy to relocate.
Amber cabinets reduce visible light penetration by more than 50% and blocks 99% of UV light.
  • Has two perforated shelves that are removable
  • Door has two latches at the top and loops for - lock or tamper evident seal
Other Notes
Model 1.0 has 1 fixed shelf and 1 door latch. Models 3.0 and 4.0 have 3 removable shelves.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)