Key Documents
Z507660
Sigma-Aldrich
Ace gas dispersion tube with porous fritted glass tip
Fit #7 Ace-Thred, O.D. × L 7 mm × 135 mm, porosity 25-50 μm (C)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
manufacturer/tradename
Ace Glass 943523
O.D. × L
7 mm × 135 mm
pore size
25-50 μm porosity (C)
Application
Ace gas dispersion tube with porous fritted glass tip is suitable for fine dispersion.
Choose from one of the most recent versions:
Certificates of Analysis (COA)