Key Documents
Z308862
Quickfit® conical flasks
capacity 500 mL, joint: ST/NS 19/26
Synonym(s):
FE500/2
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
conical flask
flat bottom
Joint
joint: ST/NS19/26
sterility
non-sterile
feature
closure type Standard Taper Joint
without cap
packaging
case of 5
manufacturer/tradename
Quickfit FE500/2
availability
not available in USA and Canada
capacity
500 mL
General description
With ground-glass socket for general use. Not suitable for vacuum use.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)